Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 v2 Rat anti-Mouse, Brilliant Violet 650, Clone: 587702, BD OptiBuild™

Catalog No. BDB747931 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB747931 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB747931 Supplier BD Biosciences Supplier No. 747931
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD97 v2 Monoclonal antibody specifically detects CD97 v2 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD97 v2
Applications Flow Cytometry
Classification Monoclonal
Clone 587702
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 650
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD97 v2
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.