Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 (4F2) Rat anti-Mouse, BUV615, Clone: H202-141, BD OptiBuild™

Catalog No. BDB752360 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752360 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752360 Supplier BD Biosciences Supplier No. 752360
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD98 (4F2) Monoclonal antibody specifically detects CD98 (4F2) in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD98 (4F2)
Applications Flow Cytometry
Classification Monoclonal
Clone H202-141
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms CD98 BUV615 H202-141 50 μg
Host Species Rat
Immunogen Not Reported
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD98 (4F2)
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.