Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD98 Antibody (HBJ127), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281109AF594
Description
CD98 Monoclonal specifically detects CD98 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CD98 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
SLC3A2 | |
HBJ127 was obtained from hybridomas generated by the fusion of mouse myeloma cells and spleen cells from mice which had been immunized with T24 hyman bladder cancer cells. | |
0.1 mL | |
Biologically Active Proteins, Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
6520 | |
Human | |
Purified |
ELISA, Immunofluorescence | |
HBJ127 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
HBJ127 binds to the CD98 heavy chain at an epitope consisting of residues AFS (aa 442-444) (Itoh et al, 2007). The antibody has not been shown to cross-react with other species. CD98 is expressed at high levels on monocytes and at very low level on peripheral blood T and B lymphocytes, splenocytes, NK cells and granulocytes. CD98 is a disulfide-linked and glycosylated type II integral membrane protein. This protein has roles in normal and neoplastic cell growth and is required for the function of light chain amino acid transporters. It is also involved in sodium-independent transport of large neitral amino acids and in guiding and targeting LAT1 and LAT2 to the plasma membrane. When associated with SLC7A6 or SLC7A7, CD98 acts as an arginine/glutamine exchanger. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction