Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD98 Mouse anti-Human, Alexa Fluor™ 647, Clone: MEM-108, BD Pharmingen™
Mouse Monoclonal Antibody
$144.00 - $348.00
Specifications
Antigen | CD98 |
---|---|
Clone | MEM-108 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
CD98 Monoclonal antibody specifically detects CD98 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD98 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Human | |
SLC3A2; CD98; CD98 Heavy Chain; CD98HC; 4F2; 4F2hc | |
IgG1 κ | |
Affinity Chromatography |
MEM-108 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Raji Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title