Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98 Mouse anti-Human, Brilliant Violet 650, Clone: UM7F8, BD Optibuild™

Catalog No. BDB744505 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744505 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744505 Supplier BD Biosciences Supplier No. 744505
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD98 is a disulfide-linked heterodimer composed of a ñ80-85 kDa, type II membrane glycoprotein heavy chain, (CD98hc), and ñ40-45 kDa light chain. The UM7F8 monoclonal antibody specifically recognizes CD98hc, which is also known as 4F2 heavy chain antigen (4F2hc). CD98 is broadly expressed on hematopoietic cells, including peripheral blood lymphocytes, monocytes and granulocytes (low), as well as non-hematopoietic cells, eg, intestinal epithelial cells. CD98 expression is upregulated on activated and proliferating cells. CD98hc is encoded by the SLC3A2 [solute carrier family 3 (amino acid transporter heavy chain), member 2] gene. CD98 reportedly functions in transmembrane amino acid transport and in the regulation of integrin signaling which are involved in the regulation of cellular activation, proliferation, and survival. The UM7F8 antibody is reportedly a functional antibody that can costimulate T cell proliferative responses.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD98
Classification Monoclonal
Clone UM7F8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SLC3A2, CD98HC, 4F2, 4F2hc, MDU1, NACAE, 4T2HC
Host Species Mouse
Immunogen Human T-leukemic and Thymic Cell Lines
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.