Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD98/SLC3A2 Monoclonal Antibody (2B10F5), CoraLite488, Proteintech
SDP

Catalog No. 502334624 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4624 Supplier Proteintech Group Inc Supplier No. CL48866883100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD98/SLC3A2 Monoclonal antibody specifically detects CD98/SLC3A2 in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen CD98/SLC3A2
Applications Immunocytochemistry
Classification Monoclonal
Clone 2B10F5
Concentration 1 mg/mL
Conjugate CoraLite 488
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. P08195
Gene Alias 4F2; 4F2HC; 4T2HC; CD98; CD98HC; MDU1; NACAE; SLC3A2
Host Species Mouse
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6520
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.