Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD99 Antibody (HO36-1.1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2446920.1MG
Description
CD99 Monoclonal antibody specifically detects CD99 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
CD99 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 5 - 10 ul/million cells in 0.1 ml, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunofluorescence 0.5 - 1.0 ug/ml | |
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
Purified E-rosette forming cells from human peripheral blood lymphocytes | |
0.1 mg | |
Immunology | |
4267 | |
Store at 4C. | |
IgM κ |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
HO36-1.1 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein L purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction