Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD99 Antibody (MIC2/3478R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308589
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
CD99 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
Synthetic peptide corresponding to CD99 residues within aa85 C terminal of CD99 was used as an immunogen. | |
100 μg | |
Immunology | |
4267 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
MIC2/3478R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction