Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD99 Antibody (MIC2/877), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247895MFV610
Description
CD99 Monoclonal antibody specifically detects CD99 in Human, Rat (Negative) samples. It is validated for Flow CytometrySpecifications
CD99 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat (Negative) | |
Purified |
Flow Cytometry | |
MIC2/877 | |
Flow Cytometry | |
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
Recombinant full-length human CD99 protein (Uniprot: P14209) | |
0.1 mL | |
Immunology | |
4267 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction