Learn More
CD99/MIC2 (Ewing' s Sarcoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: HO36-1.1
Isotype: IgM
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified E-rosette forming cells from human peripheral blood lymphocytes
Molecular Weight: 27-32kDa
Positive Control: Pancreas or Ewing's sarcoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Specifications
| Antigen | CD99/MIC2 (Ewings Sarcoma Marker) Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HO36-1.1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified E-rosette forming cells from human peripheral blood lymphocytes |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.