Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99/MIC2 (Ewing' s Sarcoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS294P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS294P1 500 μL Concentrated 200μg/mL
MS294P0 100 μL Concentrated 200μg/mL
MS294P 1 mL Concentrated 200μg/mL
MS294P1ABX 100 μL Concentrated 1mg/mL
MS294PABX 200 μL Concentrated 1mg/mL
MS294R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS294P1 Supplier Epredia Supplier No. MS294P1

Detect CD99 in human and rat samples with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-1, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.

Host Species: Mouse

Clone: HO36-1.1

Isotype: IgM

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified E-rosette forming cells from human peripheral blood lymphocytes

Molecular Weight: 27-32kDa

Positive Control: Pancreas or Ewing's sarcoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen CD99/MIC2 (Ewings Sarcoma Marker) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HO36-1.1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified E-rosette forming cells from human peripheral blood lymphocytes
Quantity 500 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.