Learn More
CD99 Mouse anti-Human, Clone: OTI2H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800821

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.Specifications
CD99 | |
Monoclonal | |
Unconjugated | |
CD99 | |
HBA71, MIC2, MIC2X, MIC2Y, MSK5X | |
Mouse | |
Affinity Chromatography | |
RUO | |
4267 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P14209 | |
CD99 | |
Full length human recombit protein of human CD99 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.