Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Mouse anti-Human, Unconjugated, Clone: 12E7 + MIC2/877, Abnova™

Catalog No. 89337781 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-781 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-781 Supplier Abnova Corporation Supplier No. MAB21290
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen CD99
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 12E7 + MIC2/877
Conjugate Unconjugated
Description CD99 molecule
Gene CD99
Gene Alias MIC2, MIC2X, MIC2Y
Host Species Mouse
Immunogen Human acute lymphocytic leukemia T-cells (12E7) and recombinant human MIC2 protein (MIC2/877)
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4267
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.