Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Antibody (MIC2/1495R), Novus Biologicals™
SDP

Catalog No. p-7078743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25336820 20 μg
NB253368100 100 μg
NB253368200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25336820 Supplier Novus Biologicals Supplier No. NBP25336820UG
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD99
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MIC2/1495R
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2 - 4ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias 12E7, antigen identified by monoclonal antibodies 12E7, F21 and O13, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
Gene Symbols CD99
Host Species Rabbit
Immunogen Recombinant human full-length MIC2 protein
Molecular Weight of Antigen 29 kDa
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4267
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.