Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD99 Rabbit anti-Human, Clone-MIC2/1495R, Novus Biologicals™

Rabbit Monoclonal Antibody
$610.00
Specifications
Antigen | CD99 |
---|---|
Clone | MIC2/1495R |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 2 - 4 ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254373100
![]() |
Novus Biologicals
NBP254373100UG |
100 μg |
Each for $610.00
|
|
|||||
NB254373200
![]() |
Novus Biologicals
NBP254373200UG |
200 μg | N/A | N/A | N/A | ||||
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
CD99 | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
RUO | |
Human | |
12E7, antigen identified by monoclonal antibodies 12E7, F21 and O13, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
CD99 | |
IgG κ | |
Protein A purified | |
29 kDa |
MIC2/1495R | |
Flow Cytometry 2 - 4 ug/million cells, Immunocytochemistry/Immunofluorescence 2 - 4 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, CyTOF-ready | |
Monoclonal | |
Rabbit | |
Immunology | |
PBS with No Preservative | |
4267 | |
Recombinant human full-length MIC2 protein | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title