Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD99 Rabbit anti-Human, Clone: MIC2/3478R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
Antigen | CD99 |
---|---|
Clone | MIC2/3478R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
CD99 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Immunology | |
10 mM PBS with 0.05% BSA | |
4267 | |
IgG | |
Protein A purified |
MIC2/3478R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
Synthetic peptide corresponding to CD99 residues within aa85 C terminal of CD99 was used as an immunogen. | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title