Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD99L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578592 Supplier Invitrogen™ Supplier No. PA578592
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Rhesus Monkey (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CD99L2 may function as a homophilic adhesion molecule. CD99L2 functions in leukocyte-endothelial cell interactions during leukocyte extravasation, and in particular, at the diapedesis step. CD99L2 does not seem to be involved in docking of leukocytes to the vessel wall or in lymphocyte diapedesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD99L2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Cd99l2
Gene Accession No. Q8TCZ2
Gene Alias AW548191; CD99; Cd99 antigen-like 2; CD99 antigen-like protein 2; CD99 molecule like 2; CD99 molecule-like 2; CD99B; CD99L2; MIC2 (monoclonal Imperial Cancer Research Fund 2)-like 1; MIC2 like 1; MIC2 like-1; MIC2L1; MIC2-like protein 1; protein rhombex-40; Rhombencephalic expression protein 40 kDa; Rhombex40; Rhombex-40; single-pass transmembrane protein; UNQ1964/PRO4486; vms-tm2; Xap89
Gene Symbols Cd99l2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CD99L2. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.