Learn More
Invitrogen™ CD99L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590808
Description
Immunogen sequence: DFDDFNLEDA VKETSSVKQP WDHTTTTTTN RPGTTRAPAK PPGSGLDLAD ALDDQDDGRR KPGIGGRERW NHVTTTTKRP VTTRAPANTL GNDFDLADAL DDRNDRDDGR RKPIAGGGGF SDKDLEDIVG GGEYKPDKGK GDGRYGSNDD PGSGMVAEPG; Positive Samples: HeLa, Mouse brain, Mouse kidney, Mouse liver, Mouse lung, Rat pancreas; Cellular Location: Cell junction, Cell membrane, Extracellular side, Single-pass type I membrane protein.
CD99L2 may function as a homophilic adhesion molecule. CD99L2 functions in leukocyte-endothelial cell interactions during leukocyte extravasation, and in particular, at the diapedesis step. CD99L2 does not seem to be involved in docking of leukocytes to the vessel wall or in lymphocyte diapedesis.
Specifications
CD99L2 | |
Polyclonal | |
Unconjugated | |
Cd99l2 | |
AW548191; CD99; Cd99 antigen-like 2; CD99 antigen-like protein 2; CD99 molecule like 2; CD99 molecule-like 2; CD99B; CD99L2; MIC2 (monoclonal Imperial Cancer Research Fund 2)-like 1; MIC2 like 1; MIC2 like-1; MIC2L1; MIC2-like protein 1; protein rhombex-40; Rhombencephalic expression protein 40 kDa; Rhombex40; Rhombex-40; single-pass transmembrane protein; UNQ1964/PRO4486; vms-tm2; Xap89 | |
Rabbit | |
Affinity chromatography | |
RUO | |
171485, 171486, 83692 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.6 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BIF0, Q8R1R5, Q8TCZ2 | |
Cd99l2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 26-185 of human CD99L2 (NP_1136502). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.