Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDA1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727702 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7702 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7702 Supplier Proteintech Group Inc Supplier No. 120872AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal domit polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.

Specifications

Antigen CDA1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TSPYL2
Gene Accession No. Q9H2G4
Gene Alias CDA1, Cell division autoantigen 1, CINAP, CTCL, DENTT, NP79, Nuclear protein of 79 kDa, TSPX, TSPYL2
Gene Symbols TSPYL2
Host Species Rabbit
Immunogen CDA1 Fusion Protein Ag2722
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64061
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.