Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDADC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568390 Supplier Invitrogen™ Supplier No. PA568390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDADC1 Polyclonal Antibody for Western Blot, IHC (P)

May play an important role in testicular development and spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDADC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CDADC1
Gene Accession No. Q8BMD5, Q9BWV3
Gene Alias 2310010M10Rik; AI449174; AI463398; bA103J18.1; BB130816; Cdadc1; cytidine and dCMP deaminase domain containing 1; cytidine and dCMP deaminase domain-containing protein 1; NYD-SP15; protein kinase NYD-SP15; testis development protein NYD-SP15
Gene Symbols CDADC1
Host Species Rabbit
Immunogen Synthetic protein corresponding to residues near the N terminal of human CDADC1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71891, 81602
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.