Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDAN1 Antibody (Cod177), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250176AF700
Description
CDAN1 Monoclonal specifically detects CDAN1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CDAN1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDAN1 | |
aa 294-607 of cloned human protein | |
0.1 mL | |
Cardiovascular Biology | |
146059 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Cod177 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CDA1, CDAI, CDA-I, codanin, codanin-1, congenital dyserythropoietic anemia, type I, discs lost homolog, DLT, PRO1295 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction