Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558776 Supplier Invitrogen™ Supplier No. PA558776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NCVKHIKATL VADLVRQAES LLQEQLVTQG EEGGDPAQLL EILCSQLCPH GAQALALGRE FCQRKSPGAV RALLPEETPA AVLSSAENIA VGLA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

This gene encodes a protein that appears to play a role in nuclear envelope integrity, possibly related to microtubule attachments. Mutations in this gene cause congenital dyserythropoietic anemia type I, a disease resulting in morphological and functional abnormalities of erythropoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDAN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDAN1
Gene Accession No. Q8IWY9
Gene Alias 1500015A01Rik; AI448026; AW492297; CDA1; CDAI; CDA-I; Cdan1; CDAN1A; codanin 1; codanin-1; congenital dyserythropoietic anemia type I; congenital dyserythropoietic anemia, type I; congenital dyserythropoietic anemia, type I (human); discs lost homolog; DLT; PRO1295; UNQ664/PRO1295
Gene Symbols CDAN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDAN1. Recombinant protein control fragment (Product #RP-98094).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146059
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.