Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987975
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7975 Supplier Bioss Supplier No. BS7741R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.

Specifications

Antigen CDC10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SEPTIN7
Gene Accession No. Q16181
Gene Alias 7-Sep; Cdc10; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; CDC3; cell division cycle 10 homolog; E430034N22; NBLA02942; Sept7; SEPT7A; septin 7; septin 7 variant 4; SEPTIN7; Septin-7
Gene Symbols SEPTIN7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Septin 7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 989
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.