Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC123 Antibody (SR1649), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CDC123 |
---|---|
Clone | SR1649 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Description
CDC123 Monoclonal antibody specifically detects CDC123 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
CDC123 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cell division cycle 123 homolog (S. cerevisiae), cell division cycle protein 123 homolog, chromosome 10 open reading frame 7, D123C10orf7, FLJ13863, HT-1080, Protein D123, PZ32 | |
A synthesized peptide derived from human CDC123 (Uniprot #: O75794) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1649 | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
8872 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title