Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDC123 Monoclonal Antibody (OTI1F8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526184
Description
CDC123 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid. The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase, and a homodimeric dihydrolipoamide dehydrogenase. E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
Specifications
CDC123 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75794 | |
CDC123 | |
Full length human recombinant protein of CDC123 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Canine, Monkey | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI1F8 | |
Unconjugated | |
CDC123 | |
AA959893; C10orf7; Cdc123; cell division cycle 123; cell division cycle 123 homolog; cell division cycle protein 123 homolog; Cell division cycle protein 123-like protein; D123; D123 gene product; G431001I09Rik; HT-1080; hypothetical protein; Protein D123; PZ32; QccE-12339 | |
Mouse | |
Affinity chromatography | |
RUO | |
478005, 8872 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction