Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114232 Supplier Invitrogen™ Supplier No. PA5114232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDDVEMKNGI TQGDKLRALK SQRQPRTSPS CAFRSDDTKG HPRAVSQPFR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 87%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%.

The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc14A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDC14A
Gene Accession No. Q9UNH5
Gene Alias A830059A17Rik; CDC10 (cell division cycle 10, S. cerevisiae, homolog); cdc14; CDC14 cell division cycle 14 homolog A; CDC14 cell division cycle 14A; CDC14A; CDC14a1; CDC14A2; cell division cycle 14A; DFNB105; Dual specificity protein phosphatase CDC14A; hCDC14
Gene Symbols CDC14A
Host Species Rabbit
Immunogen Immunogen sequence: DDDVEMKNGI TQGDKLRALK SQRQPRTSPS CAFRSDDTKG HPRAVSQPFR (aa 384-433).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8556
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.