Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDC14B/CDC14C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599828 Supplier Invitrogen™ Supplier No. PA599828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDC14B/C.

The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. This protein is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, which suggests the role in cell cycle control. This protein has been shown to interact with and dephosphorylates tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splice of this gene results in 3 transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC14B/CDC14C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cdc14b
Gene Accession No. A4D256, O60729, Q6PFY9
Gene Alias 2810432N10Rik; A530086E13Rik; AA472821; CDC14 cell division cycle 14 homolog B; CDC14 cell division cycle 14B; CDC14B; Cdc14B1; Cdc14B2; CDC14B3; CDC14Bretro; CDC14C; cell division cycle 14B; cell division cycle 14C; dual specificity protein phosphatase CDC14B; hCDC14B
Gene Symbols Cdc14b, CDC14C
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDC14B(Accession O60729), corresponding to amino acid residues A14-R64.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168448, 218294, 8555
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.