Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc14B Polyclonal Antibody
GREENER_CHOICE

Catalog No. 348900
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-890-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-890-0 Supplier Invitrogen™ Supplier No. 348900
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Cdc14B Polyclonal Antibody for Western Blot, ICC/IF, IP

CDC14B is a member of the dual specificity protein tyrosine phosphatase family. This protein is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, which suggests the role in cell cycle control. This protein has been shown to interact with and dephosphorylates tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splice of this gene results in 3 transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc14B
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CDC14B
Gene Accession No. O60729, Q6PFY9
Gene Alias 2810432N10Rik; A530086E13Rik; AA472821; CDC14 cell division cycle 14 homolog B; CDC14 cell division cycle 14B; CDC14B; Cdc14B1; Cdc14B2; CDC14B3; cell division cycle 14B; dual specificity protein phosphatase CDC14B; hCDC14B
Gene Symbols CDC14B
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of the human Cdc14B protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218294, 8555
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.