Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC2/CDK1 Antibody (CDK1/873), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311507G
Description
CDC2/CDK1 Monoclonal antibody specifically detects CDC2/CDK1 in Human, Bovine, Mink, Monkey, S. pombe (Negative), Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative), Yeast (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CDC2/CDK1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Mink, Monkey, S. pombe (Negative), Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative) | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CDK1/873 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2 | |
Recombinant human full-length CDC2/CDK1 protein. (Uniprot: P06493) | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
983 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction