Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals CDC2/CDK1 Antibody (POH-1; same as cdc2.1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2347110.1MG
Description
CDC2/CDK1 Monoclonal antibody specifically detects CDC2/CDK1 in Human, Bovine, Mink, Monkey, Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOFSpecifications
CDC2/CDK1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 0.5-1μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready | |
CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2 | |
Recombinant full-length human p34cdc2 protein (Uniprot: P06493) | |
0.1 mg | |
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
983 | |
Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
POH-1; same as cdc2.1 | |
Unconjugated | |
10 mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Mink, Monkey, Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction