Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2 p34Rabbit anti-Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501990028
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0028 Supplier Rockland Immunochemicals Supplier No. 100401160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Antiserum will specifically react with a 34 kDa cdc2 protein from human, rat and mouse tissue. No reaction was observed against other related cyclin dependent kinases. Cross reactivity with cdc2 from other species may also occur.

CDK1 (CDC2, P34CDC2) is a member of the Ser/Thr protein kinase family. CDK1 is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of CDK1 is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of CDK1 also play important regulatory roles in cell cycle control. Destruction of cyclin B during metaphase results in inactivation of CDK1, allowing mitotic exit and cell division. CDK1 also contributes to the control of DNA replication and can be inhibited by several transcriptional targets of p53, such as p21WAF. The ability of CDK1 to exercise control over the cell cycle is dependent upon the phosphorylation of Tyr15 in cdc2. Research studies have shown that expression of CDK1 in the brain has been linked to both neurogenesis and apoptosis. In vertebrates, multiple cyclins like CDK1 have been identified, including a total of eight such regulatory proteins in mammals. Alternatively spliced transcript variants encoding different isoforms of CDK1 have been found.

Specifications

Antigen CDC2 p34
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.01% sodium azide; pH
Gene CDK1
Gene Accession No. P06493, P11440, P39951
Gene Alias 5363; ABL1; BOS_24079; cdc; CDC2; cdc-2; cdc2 kinase; CDC28A; Cdc2a; cdc2-a; cdc2Dm; cdc2x1.1; CDCDm; CDK1; Cdk-1; cdk1.S; CDK1/CDC2; cdk1-a; cdk1-b; Cdk1-PA; CDKN1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control; Cell division control protein 2 homolog; Cell division control protein 2 homolog 1; Cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; Cell division protein kinase 1; Cell division protein kinase 1-A; CG5363; CG5363 gene product from transcript CG5363-RA; CG5363-PA; cyclin dependent kinase; cyclin dependent kinase 1; cyclin-dependent kinase; Cyclin-dependent kinase 1; cyclin-dependent kinase 1 S homeolog; Cyclin-dependent kinase 1-A; D2366; Dcdc2; dCdk1; DKFZp686L20222; Dm cdc2; Dmcdc2; DmCdk1; Dmel\CG5363; Dmel_CG5363; DNA polymerase delta catalytic subunit; DNA polymerase III; DNA-directed DNA polymerase delta POL3; group 4; HPR6; KCIP-1; l(2)31Eh; maturation-promoting factor; MGC111195; MGC53829; MPF; OTTHUMP00000019660; P34 CDK1; P34 MGC111195; p34 protein kinase; p34 protein kinase 1; p34; P34CDC2; POL3; Protein 1054; protein 2; PSTAIR; TEX1; xcdc2; XELAEV_18036802mg; YDL102W
Gene Symbols CDK1
Host Species Rabbit
Immunogen CDC2 peptide corresponding to the C-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12534, 54237, 983
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.