Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC2 Rabbit anti-Human, Unconjugated, Clone: BOI-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19875
Description
CDC2 Monoclonal antibody specifically detects CDC2 in Human samples. It is validated for WB-Ce.Specifications
CDC2 | |
Monoclonal | |
Unconjugated | |
CDC2 | |
Rabbit | |
100 μg | |
983 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
BOI-3 | |
cell division cycle 2, G1 to S and G2 to M | |
CDC28A, CDK1, DKFZp686L20222, MGC111195 | |
A synthetic peptide corresponding to human CDC2 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction