Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc20 Antibody (BA8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002646F
Description
Cdc20 Monoclonal specifically detects Cdc20 in Xenopus samples. It is validated for Flow Cytometry, ELISA.Specifications
Cdc20 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CDC20 | |
Peptide from the carboxy terminus of the protein. | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
991 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, ELISA | |
BA8 | |
Flow Cytometry, ELISA | |
CDC20 cell division cycle 20 homolog, CDC20 cell division cycle 20 homolog (S. cerevisiae), cell division cycle 20 homolog (S. cerevisiae), cell division cycle protein 20 homolog, MGC102824, p55CDCS. cerevisiae, homolog) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction