Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563103 Supplier Invitrogen™ Supplier No. PA563103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPTKKEHQKA WALNLNGFDV EEAKILRLSG KPQNAPEGYQ NRLKVLYSQK ATPGSSRKTC RYIPSLPDRI LDAPEIRNDY Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.

Required for full ubiquitin ligase activity of the anaphase promoting complex/cyclosome (APC/C) and may confer substrate specificity upon the complex. Is regulated by MAD2L1: in metaphase the MAD2L1-CDC20-APC/C ternary complex is inactive and in anaphase the CDC20-APC/C binary complex is active in degrading substrates. The CDC20-APC/C complex positively regulates the formation of synaptic vesicle clustering at active zone to the presynaptic membrane in postmitotic neurons. CDC20-APC/C-induced degradation of NEUROD2 induces presynaptic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc20
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC20
Gene Accession No. Q12834
Gene Alias 2310042N09Rik; bA276H19.3; C87100; cdc20; CDC20 cell division cycle 20 homolog; cdc20.S; CDC20A; cell cycle protein p55CDC; cell division cycle 20; cell division cycle 20 homolog; cell division cycle 20 S homeolog; cell division cycle protein 20 homolog; fizzy1; mmCdc20; p55cdc; XELAEV_18025163mg; X-FZY
Gene Symbols CDC20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cdc20. Recombinant protein control fragment (Product #RP-103211).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 991
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.