Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC20B Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728692 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-172-8692 150 μL
1 options

Catalog No. 50-172-8692

Supplier: Proteintech Group Inc 133761AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CDC20B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CDC20B
Gene Accession No. Q86Y33
Gene Alias CDC20B, G6VTS76519
Gene Symbols CDC20B
Host Species Rabbit
Immunogen CDC20B Fusion Protein Ag4198
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166979
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.