Learn More
Invitrogen™ Cdc23 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-20949 can be used with blocking peptide PEP-1063.
Specifications
Specifications
| Antigen | Cdc23 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Cdc23 |
| Gene Accession No. | Q8BGZ4, Q9UJX2 |
| Gene Alias | 6030435O18; ANAPC8; anaphase promoting complex subunit 8; anaphase-promoting complex subunit 8; APC8; CDC23; CDC23 (cell division cycle 23, yeast, homolog); CDC23 cell division cycle 23; cell division cycle 23; cell division cycle 23 homolog; cell division cycle protein 23; cell division cycle protein 23 homolog; CUT23; cyclosome subunit 8; D18Ertd243e; zgc:56013 |
| Gene Symbols | Cdc23 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.