Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC25A Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100213B
Description
CDC25A Polyclonal antibody specifically detects CDC25A in Human samples. It is validated for Western BlotSpecifications
CDC25A | |
Polyclonal | |
Western Blot | |
CDC25A2, CDC25A2-CAG isoform, cell division cycle 25 homolog A (S. cerevisiae), cell division cycle 25 homolog A (S. pombe), cell division cycle 25A, Dual specificity phosphatase Cdc25A, EC 3.1.3.48, M-phase inducer phosphatase 1 | |
The immunogen recognized by this antibody maps to a region between residues 175 and 225 of human Cell Division Cycle 25A using the numbering given in entry NP_001780.2 (GeneID 993). | |
0.1 mL | |
Adaptive Immunity, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase | |
993 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction