Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc25A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577902 Supplier Invitrogen™ Supplier No. PA577902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: H1299, MCF-7 Predicted Reactivity: Dog (89%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CDC25a is a member of the CDC25 family of phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. CDC25a is required for progression through G1 and S phases of the cell cycle. CDC25a activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division. CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc25A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CDC25A
Gene Accession No. P30304, P48964, P48965
Gene Alias CDC25 isoform A1-CAG; CDC25A; CDC25A2; CDC25A2-CAG isoform; Cdc25m3; cell division cycle 25A; D9Ertd393e; dual specificity phosphatase Cdc25A; EC 3.1.3.48; M-phase inducer phosphatase 1; MPIP1
Gene Symbols CDC25A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human CDC25A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12530, 171102, 993
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.