Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC25B Monoclonal Antibody (OTI11C5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812342


Description
CDC25B Monoclonal antibody specifically detects CDC25B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CDC25B | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8CHK4, Q92993, Q99MK2 | |
Mouse | |
Protein A/G | |
RUO | |
10524, 192218, 81601 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI11C5 | |
Unconjugated | |
KAT5 | |
AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2 | |
Human recombinant protein fragment corresponding to amino acids 227-504 of human KAT5 (NP_006379) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction