Learn More
Invitrogen™ CDC25B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587450
Description
Immunogen sequence: MEVPQPEPAP GSALSPAGVC GGAQRPGHLP GLLLGSHGLL GSPVRAAASS PVTTLTQTMH DLAGLGSETP KSQVGTLLFR SRSRLTHLSL SRRASESSLS SESSESSDAG LCMDSPSPMD PHMAEQTFEQ AIQAASRIIR NEQFAIRRFQ SMPVRLLGHS PVLRNITNSQ APDGRRKSEA GSGAASSSGE DKENDGFVFK MPWKPTHPSS THALAEWASR REAFAQRPSS APDLMCLSPD RKMEVEELSP; Positive Samples: A-549; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle pole.
Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.
Specifications
CDC25B | |
Polyclonal | |
Unconjugated | |
CDC25B | |
AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
994 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.93 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P30305 | |
CDC25B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human CDC25B (NP_0686591). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.