Learn More
Invitrogen™ Cdc25B Polyclonal Antibody

Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | Cdc25B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 278 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100 μg/mL BSA, 0.15M NaCl, 50% glycerol and no preservative; pH 7.5 |
| Gene | CDC25B |
| Gene Accession No. | P30305, P30306, P48966 |
| Gene Alias | AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2 |
| Gene Symbols | CDC25B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.