Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDC25B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579015
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579015 Supplier Invitrogen™ Supplier No. PA579015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Kidney Tissue, Rat Lung Tissue. IHC: human lung cancer tissue.

Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC25B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene CDC25B
Gene Accession No. P30305, P48966
Gene Alias AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2
Gene Symbols CDC25B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Cdc25B (541-559aa DYRPMNHEAFKDELKTFRL).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171103, 994
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.