Learn More
Invitrogen™ CDC25B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579016
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell lysates, human Raji whole cell lysates, human Hela whole cell lysates, human HepG2 whole cell lysates, rat liver tissue lysates, mouse liver tissue lysates. IHC: human stomach cancer tissue. Flow: HepG2 cell.
Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.
Specifications
CDC25B | |
Polyclonal | |
Unconjugated | |
CDC25B | |
AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12531, 171103, 994 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P30305, P30306, P48966 | |
CDC25B | |
E.coli-derived human Cdc25B recombinant protein (Position: M119-L248). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.