Learn More
Invitrogen™ Cdc25C Monoclonal Antibody (1F12)

Description
MA5-15613 targets CDC25C in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15613 immunogen is purified recombinant fragment of human CDC25C expressed in E. Coli.. MA5-15613 detects CDC25C which has a predicted molecular weight of approximately 58kDa.
Specifications
Specifications
| Antigen | Cdc25C |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 1F12 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | Cdc25c |
| Gene Accession No. | P30307 |
| Gene Alias | CDC25; CDC25C; Cdc25m1; cell division cycle 25C; dual specificity phosphatase CDC25C; EC 3.1.3.48; mitosis inducer CDC25; M-phase inducer phosphatase 3; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; PPP1R60; protein phosphatase 1, regulatory subunit 60 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.