Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc25C Monoclonal Antibody (25C14 (TC-14)), Invitrogen™

Catalog No. PIMA514107
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514107 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514107 Supplier Thermo Scientific Supplier No. MA514107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14107 targets CDC25C in IF, IP, ChIP and WB applications and shows reactivity with Human samples. The MA5-14107 immunogen is recombinant human CDC25C protein.

M-phase inducer phosphatase 3is an enzyme that in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc25C
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 25C14 (TC-14)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Cdc25c
Gene Accession No. P30307
Gene Alias CDC25; CDC25C; Cdc25m1; cell division cycle 25C; dual specificity phosphatase CDC25C; EC 3.1.3.48; mitosis inducer CDC25; M-phase inducer phosphatase 3; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; PPP1R60; protein phosphatase 1, regulatory subunit 60
Gene Symbols Cdc25c
Host Species Mouse
Immunogen Recombinant human CDC25C protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 995
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.