Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cdc25C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595834
Description
Immunogen sequence: SFRSNQRKML NLLLERDTSF TVCPDVPRTP VGKFLGDSAN LSILSGGTPK RCLDLSNLSS GEITATQLTT SADLDETGHL DSSGLQEVHL AGMNHDQHLM KCSPAQLLCS T Positive Samples: K-562, HepG2, Raji; Cellular Location: Nucleus.
M-phase inducer phosphatase 3is an enzyme that in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.
Specifications
Cdc25C | |
Polyclonal | |
Unconjugated | |
Cdc25c | |
CDC25; CDC25C; Cdc25m1; cell division cycle 25C; dual specificity phosphatase CDC25C; EC 3.1.3.48; mitosis inducer CDC25; M-phase inducer phosphatase 3; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; PPP1R60; protein phosphatase 1, regulatory subunit 60 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12532, 307511, 995 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.72 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P30307, P48967 | |
Cdc25c | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-130 of human CDC25C (NP_0017812). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction