Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDC25C Rabbit anti-Human, Mouse, Rat, Clone: 6R0I6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CDC25C |
---|---|
Clone | 6R0I6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CDC25C Monoclonal antibody specifically detects CDC25C in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CDC25C | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Protein Phosphatase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
995 | |
IgG | |
Affinity purified |
6R0I6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CDC 25, CDC25, cell division cycle 25 homolog C (S. cerevisiae), cell division cycle 25 homolog C (S. pombe), cell division cycle 25C, Dual specificity phosphatase Cdc25C, EC 3.1.3.48, mitosis inducer CDC25, M-phase inducer phosphatase 3, phosphotyrosine phosphatase | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDC25C (P30307). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title