Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc26 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522141

Catalog No. PIPA522141


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299. Predicted reactivity: Mouse (88%), Rat (89%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CDC26, also named as ANAPC12, APC12 and C9orf17, is a component of the anaphase promoting complex/cyclosome (APC/C).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cdc26
Polyclonal
Unconjugated
CDC26
2010012C09Rik; ANAPC12; anaphase-promoting complex subunit 12; Anaphase-promoting complex subunit CDC26; APC12; BWK-2; C9orf17; CDC26; CDC26 subunit of anaphase promoting complex; cell division cycle 26; cell division cycle 26 homolog; cell division cycle protein 26 homolog; Protein BWK-2
Rabbit
Antigen affinity chromatography
RUO
246184
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8NHZ8
CDC26
Full length human CDC26 Recombinant protein.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.