Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560623
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560623 Supplier Invitrogen™ Supplier No. PA560623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLRRKPTRLE LKLDDIEEFE NIRKDLETRK KQKEDVEVVG GSDGEGAIGL SSDPKSREQM INDRIGYKPQ PKPNNRSSQF GSL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

CDC26, also named as ANAPC12, APC12 and C9orf17, is a component of the anaphase promoting complex/cyclosome (APC/C).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc26
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC26
Gene Accession No. Q8NHZ8
Gene Alias 2010012C09Rik; ANAPC12; anaphase-promoting complex subunit 12; Anaphase-promoting complex subunit CDC26; APC12; BWK-2; C9orf17; CDC26; CDC26 subunit of anaphase promoting complex; cell division cycle 26; cell division cycle 26 homolog; cell division cycle protein 26 homolog; Protein BWK-2
Gene Symbols CDC26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cdc26. Recombinant protein control fragment (Product #RP-100238).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.