Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc27 Antibody (AF 3.1), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002606AF488
Description
Cdc27 Monoclonal specifically detects Cdc27 in Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
Cdc27 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDC27 | |
A peptide corresponding to the C-terminal 10 residues of human cdc27 coupled to KLH. | |
0.1 mL | |
Cell Cycle and Replication | |
996 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
AF 3.1 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Mammalia, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction